Learn More
Cayman Chemical D-LucIferIn 50mg
SDP
Catalog No. 502932834
Encompass_Preferred
A chemiluminescent substrate of firefly luciferase which produces light upon oxidative decarboxylation in the presence of ATP
Catalog No. 50-293-2834 Supplier Cayman Chemical Supplier No. 2583650 MG
A chemiluminescent substrate of firefly luciferase which produces light upon oxidative decarboxylation in the presence of ATP