Learn More
Apexbio Technology LLC D-Luciferin(Synonyms: Firefly Luciferin, Luciferin, Beetle Luciferin, (S)-2-(6'-Hydroxybenzothiazolyl)-Δ2-thiazoline-4-carboxylic acid), 5mg, CAS: 2591-17-5.
SDP
Catalog No. 5000060424
Encompass_Preferred
D-Luciferin (CAS 2591-17-5) is a membrane-permeable bioluminescent substrate for firefly luciferase characterized by a Km of approximately 2 M In the presence of ATP luciferin undergoes oxidation and decarboxylation catalyzed by luciferase emitting measurable photons This bioluminescence enables quantification of intracellular ATP levels both in vitro and in vivo In biomedical research D-Luciferin is commonly applied for monitoring promoter-driven luciferase gene expression and as a non-invasive imaging probe in bioluminescence imaging (BLI) studies to assess tumor burden pharmacodynamics and cellular ATP content
Catalog No. 50-000-60424 Supplier Apexbio Technology LLC Supplier No. B60405
D-Luciferin (CAS 2591-17-5) is a membrane-permeable bioluminescent substrate for firefly luciferase characterized by a Km of approximately 2 M In the presence of ATP luciferin undergoes oxidation and decarboxylation catalyzed by luciferase emitting measurable photons This bioluminescence enables quantification of intracellular ATP levels both in vitro and in vivo In biomedical research D-Luciferin is commonly applied for monitoring promoter-driven luciferase gene expression and as a non-invasive imaging probe in bioluminescence imaging (BLI) studies to assess tumor burden pharmacodynamics and cellular ATP content