Learn More
Medchemexpress LLC D-Luciferin 6 -methy 100mg | 24404-90-8 | 294.35 g/mol | C12H10N2O3S2 | 100 MG
SDP
Catalog No. 5000228892
Encompass_Preferred
D-Luciferin 6′-methyl ether is a D-luciferin analog that functions as a potent inhibitor of firefly (Photinus pyralis) luciferase. It is supplied as a high-purity solid for use in biochemical and enzymatic assays to study luciferase activity and inhibitor interactions.
  • Potent firefly luciferase inhibitor with reported IC50 of approximately 0.1 μM.
  • High purity (>99.5%) suitable for biochemical assays.
  • Molecular formula C12H10N2O3S2 and molecular weight 294.35 g/mol.
  • Useful for luciferase assay development and inhibitor characterization.
  • Available in a 100 mg pack for laboratory use.
Catalog No. 50-002-28892 Supplier Medchemexpress LLC Supplier No. HY115749100MG
D-Luciferin 6′-methyl ether is a D-luciferin analog that functions as a potent inhibitor of firefly (Photinus pyralis) luciferase. It is supplied as a high-purity solid for use in biochemical and enzymatic assays to study luciferase activity and inhibitor interactions.
  • Potent firefly luciferase inhibitor with reported IC50 of approximately 0.1 μM.
  • High purity (>99.5%) suitable for biochemical assays.
  • Molecular formula C12H10N2O3S2 and molecular weight 294.35 g/mol.
  • Useful for luciferase assay development and inhibitor characterization.
  • Available in a 100 mg pack for laboratory use.