Learn More
Medchemexpress LLC D-Luciferin 6 -methy 10mg | 24404-90-8 | 294.35 g/mol | C12H10N2O3S2 | 10 MG
SDP
Catalog No. 5000308211
Encompass_Preferred
D-luciferin 6′-methyl ether is a D-luciferin analogue that acts as a potent inhibitor of Photinus pyralis luciferase (PpyLuc). It is provided as a solid reagent for biochemical and cell-based assays to probe luciferase activity and inhibitor interactions. Typical reported purity is ~99.5% and the molecular weight is 294.35 g/mol.
  • Potent PpyLuc inhibitor with reported IC50 around 0.1 μM.
  • Suitable for biochemical and cell-based assay applications.
  • High reported purity for reliable experimental results.
  • Stable when stored protected from light at 4°C; in solvent store at -80°C for long term.
  • For research use only; not for human or clinical use.
Catalog No. 50-003-08211 Supplier Medchemexpress LLC Supplier No. HY11574910MG
D-luciferin 6′-methyl ether is a D-luciferin analogue that acts as a potent inhibitor of Photinus pyralis luciferase (PpyLuc). It is provided as a solid reagent for biochemical and cell-based assays to probe luciferase activity and inhibitor interactions. Typical reported purity is ~99.5% and the molecular weight is 294.35 g/mol.
  • Potent PpyLuc inhibitor with reported IC50 around 0.1 μM.
  • Suitable for biochemical and cell-based assay applications.
  • High reported purity for reliable experimental results.
  • Stable when stored protected from light at 4°C; in solvent store at -80°C for long term.
  • For research use only; not for human or clinical use.