Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-Luciferin 6 -methy 10mM 1mL | 24404-90-8 | 294.35 g/mol | C12H10N2O3S2 | 1 ML
SDP

Catalog No. 5000228894
Encompass_Preferred

D-Luciferin 6'-methyl ether is a synthetic luciferin derivative used as a firefly luciferase inhibitor and biochemical reagent for bioluminescence assays. It is supplied as a solid or as a pre-made 10 mM solution in DMSO for convenient use in inhibitor studies and assay development.

  • Available as solid (multiple mg sizes) or 10 mM solution in DMSO, 1 mL.
  • High purity (99.55%) suitable for biochemical assays.
  • Molecular weight 294.35 g/mol; formula C12H10N2O3S2.
  • Used to study luciferase activity and inhibition in bioluminescent assays.
  • Soluble in DMSO; follow supplier storage recommendations.

Catalog No. 50-002-28894 Supplier Medchemexpress LLC Supplier No. HY11574910MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-Luciferin 6'-methyl ether is a synthetic luciferin derivative used as a firefly luciferase inhibitor and biochemical reagent for bioluminescence assays. It is supplied as a solid or as a pre-made 10 mM solution in DMSO for convenient use in inhibitor studies and assay development.

  • Available as solid (multiple mg sizes) or 10 mM solution in DMSO, 1 mL.
  • High purity (99.55%) suitable for biochemical assays.
  • Molecular weight 294.35 g/mol; formula C12H10N2O3S2.
  • Used to study luciferase activity and inhibition in bioluminescent assays.
  • Soluble in DMSO; follow supplier storage recommendations.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.