Learn More
Medchemexpress LLC D-Luciferin 6 -methy 25mg | 24404-90-8 | 294.35 g/mol | C12H10N2O3S2 | 25 MG
SDP
Catalog No. 5000308122
Encompass_Preferred
D-Luciferin 6′-methyl ether is a D-luciferin analog that acts as a potent inhibitor of Photinus pyralis (firefly) luciferase and is used in biochemical assays to study luciferase activity and inhibitor interactions. It is supplied as an off-white to light yellow solid and is typically handled in DMSO for assay preparation.
  • Potent PpyLuc inhibitor (IC50 ~0.1 μM).
  • High purity (99.55%).
  • Provided as a 25 mg solid.
  • Soluble in DMSO for assay preparation.
  • Suitable for luciferase inhibition assays and biochemical research.
Catalog No. 50-003-08122 Supplier Medchemexpress LLC Supplier No. HY11574925MG
D-Luciferin 6′-methyl ether is a D-luciferin analog that acts as a potent inhibitor of Photinus pyralis (firefly) luciferase and is used in biochemical assays to study luciferase activity and inhibitor interactions. It is supplied as an off-white to light yellow solid and is typically handled in DMSO for assay preparation.
  • Potent PpyLuc inhibitor (IC50 ~0.1 μM).
  • High purity (99.55%).
  • Provided as a 25 mg solid.
  • Soluble in DMSO for assay preparation.
  • Suitable for luciferase inhibition assays and biochemical research.