Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-Luciferin 6 -methy 50mg | 24404-90-8 | 294.35 g/mol | C12H10N2O3S2 | 50 MG
SDP

Catalog No. 5000229230
Encompass_Preferred

D-Luciferin 6'-methyl ether is a synthetic analog of D-luciferin that acts as a firefly luciferase inhibitor and is used in biochemical assays to study luciferase activity and inhibitor profiling. It is typically supplied as a solid with high purity and characterized by a molecular weight of approximately 294.35 g/mol.

  • Acts as a firefly luciferase inhibitor.
  • Suitable for luciferase inhibition and biochemical assays.
  • Supplied as an off-white to light yellow solid.
  • Molecular weight 294.35 g/mol.
  • Purity typically greater than 95%.

Catalog No. 50-002-29230 Supplier Medchemexpress LLC Supplier No. HY11574950MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-Luciferin 6'-methyl ether is a synthetic analog of D-luciferin that acts as a firefly luciferase inhibitor and is used in biochemical assays to study luciferase activity and inhibitor profiling. It is typically supplied as a solid with high purity and characterized by a molecular weight of approximately 294.35 g/mol.

  • Acts as a firefly luciferase inhibitor.
  • Suitable for luciferase inhibition and biochemical assays.
  • Supplied as an off-white to light yellow solid.
  • Molecular weight 294.35 g/mol.
  • Purity typically greater than 95%.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.