Learn More
Abcam D-Luciferin, chemiluminescent luciferase substrate, 50MG
Catalog No. 502706096
Encompass_Preferred
MW 280.3 Da, Purity >98%. Cell-permeable chemiluminescent luciferase substrate. Emits lights upon oxidative decarboxylation in the presence of ATP. Provides a bioluminescent signal for cellular ATP levels in vivo. Water soluble analogs are D-Luciferin potassium salt (ab143655) and D-Luciferin sodium salt (ab145164).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6096 Supplier Abcam Supplier No. AB14365450MG
MW 280.3 Da, Purity >98%. Cell-permeable chemiluminescent luciferase substrate. Emits lights upon oxidative decarboxylation in the presence of ATP. Provides a bioluminescent signal for cellular ATP levels in vivo. Water soluble analogs are D-Luciferin potassium salt (ab143655) and D-Luciferin sodium salt (ab145164).
The product is subject to the following: Abcam Restricted Use Statement