Learn More
Abcam D-Luciferin, ethyl ester, 10MG
Supplier:  Abcam AB27548910MG
Encompass_Preferred
MW 308.38 g/mol, Purity >97.5%. Membrane permeable analog of common firefly luciferase substrate producing 30% higher light intensity. Extremely useful for in vivo analysis of cloned luciferase activity in whole animal studies.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-512-93