Learn More
Carbosynth LLC. D-Luciferin Firefly, 2591-17-5, MFCD00042929, 1 g
SDP
Supplier:  Carbosynth LLC. FL086071G
Encompass_Preferred
Formula: C11H8N2O3S2, Molecular weight: 280.33 g/mol, Store at -15°C, Synonyms: (S)-4,5-Dihydro-2-(6-hydroxy-2-benzothiazolyl)-4-thiazolecarboxylic acid.
Catalog No. NC0803045