Learn More
Alkali Scientific D Luciferin Free Acid [4, 5 Dihydro 2 (6 hydroxy 2 benzothiazoyl) 4 thiazole carboxylic acid], 100 Mg
SDP
Catalog No. 5000661849
Encompass_Preferred
D-Luciferin Free Acid is a bioluminescence substrate formulated for high-sensitivity detection in a range of biochemical assays. When oxidized by luciferase, D-Luciferin emits light, making it a valuable tool for detecting ATP levels, measuring cellular activity, or assessing enzyme activity. This 100mg product is designed for use in laboratories where precise and sensitive measurements are required. It is commonly used in applications involving molecular biology, microbiology, and cellular research, providing excellent luminescence signals for quantification and assay development in high-throughput systems.
Catalog No. 50-006-61849 Supplier Alkali Scientific Supplier No. DLU008CP
D-Luciferin Free Acid is a bioluminescence substrate formulated for high-sensitivity detection in a range of biochemical assays. When oxidized by luciferase, D-Luciferin emits light, making it a valuable tool for detecting ATP levels, measuring cellular activity, or assessing enzyme activity. This 100mg product is designed for use in laboratories where precise and sensitive measurements are required. It is commonly used in applications involving molecular biology, microbiology, and cellular research, providing excellent luminescence signals for quantification and assay development in high-throughput systems.