Learn More
Alkali Scientific D Luciferin Free Acid [4, 5 Dihydro 2 (6 hydroxy 2 benzothiazoyl) 4 thiazole carboxylic acid], 250 Mg
SDP
Catalog No. 5000660338
Encompass_Preferred
D-Luciferin Free Acid (250mg) is a bioluminescence substrate used for high-sensitivity detection in various biochemical applications. The substrate, upon oxidation by luciferase, produces light, enabling the quantification of ATP and the monitoring of cellular activities in research and clinical laboratories. This form of D-Luciferin is ideal for applications requiring stable light output for sensitive detection assays. Commonly used in molecular biology, pharmacology, and environmental monitoring, this high-quality reagent ensures optimal performance for light emission detection and enhanced assay sensitivity in critical biological studies.
Catalog No. 50-006-60338 Supplier Alkali Scientific Supplier No. DLU009CP
D-Luciferin Free Acid (250mg) is a bioluminescence substrate used for high-sensitivity detection in various biochemical applications. The substrate, upon oxidation by luciferase, produces light, enabling the quantification of ATP and the monitoring of cellular activities in research and clinical laboratories. This form of D-Luciferin is ideal for applications requiring stable light output for sensitive detection assays. Commonly used in molecular biology, pharmacology, and environmental monitoring, this high-quality reagent ensures optimal performance for light emission detection and enhanced assay sensitivity in critical biological studies.