Learn More
Alkali Scientific D Luciferin Free Acid [4, 5 Dihydro 2 (6 hydroxy 2 benzothiazoyl) 4 thiazole carboxylic acid], 50 Mg
SDP
Catalog No. 5000660984
Encompass_Preferred
D-Luciferin Free Acid is a bioluminescence substrate used for high-sensitivity detection in biochemical assays. This reagent contains D-Luciferin, a substrate that is oxidized by the enzyme luciferase, resulting in light emission that can be measured for various applications. The free acid form is commonly used in bioluminescence assays for detecting ATP or monitoring cellular activity in research and clinical settings. This 50mg product is ideal for use in laboratories requiring high sensitivity for low-light detection, providing reliable and accurate results in a variety of biological assays and experimental setups.
Catalog No. 50-006-60984 Supplier Alkali Scientific Supplier No. DLU007CP
D-Luciferin Free Acid is a bioluminescence substrate used for high-sensitivity detection in biochemical assays. This reagent contains D-Luciferin, a substrate that is oxidized by the enzyme luciferase, resulting in light emission that can be measured for various applications. The free acid form is commonly used in bioluminescence assays for detecting ATP or monitoring cellular activity in research and clinical settings. This 50mg product is ideal for use in laboratories requiring high sensitivity for low-light detection, providing reliable and accurate results in a variety of biological assays and experimental setups.