Learn More
Alkali Scientific D Luciferin Free Acid [4, 5 Dihydro 2 (6 hydroxy 2 benzothiazoyl) 4 thiazole carboxylic acid], 500 Milligrams
SDP
Catalog No. 5000660986
Encompass_Preferred
D-Luciferin Free Acid is a high-purity biochemical reagent used in various laboratory applications, particularly in bioluminescence assays. It plays a crucial role in luciferase-based detection systems, where it acts as a substrate to produce light when catalyzed by luciferase enzymes. This reagent is used in a variety of applications such as biological research, molecular biology, and bioassays. The 500mg pack is perfect for laboratory use, ensuring precise and reliable results. It is essential for research in areas like cell signaling, gene expression studies, and protein quantification assays.
Catalog No. 50-006-60986 Supplier Alkali Scientific Supplier No. DLU015CP
D-Luciferin Free Acid is a high-purity biochemical reagent used in various laboratory applications, particularly in bioluminescence assays. It plays a crucial role in luciferase-based detection systems, where it acts as a substrate to produce light when catalyzed by luciferase enzymes. This reagent is used in a variety of applications such as biological research, molecular biology, and bioassays. The 500mg pack is perfect for laboratory use, ensuring precise and reliable results. It is essential for research in areas like cell signaling, gene expression studies, and protein quantification assays.