Learn More
Research Products International Corp D-Luciferin Free Acid 500 MG
SDP
Catalog No. 502230425
Encompass_Preferred
Used with firefly luciferase in bioluminescence for the determination of ATP.
Catalog No. 50-223-0425 Supplier Research Products International Corp Supplier No. L370000.5
Used with firefly luciferase in bioluminescence for the determination of ATP.