Learn More
Research Products International Corp D-Luciferin Free Acid [4,-5-Dihydro-2-(6-hydroxy-2-benzothiazoyl)-4-thiazole-carboxylic acid], 50 Milligrams
SDP
Supplier:  Research Products International Corp L370000.05
Encompass_Preferred
Used with firefly luciferase in bioluminescence for the determination of ATP.
Catalog No. 50-213-408