Learn More
Carbosynth LLC. D-Luciferin Firefly, potassium salt, 115144-35-9, MFCD0044928, 0.25 g
SDP
Catalog No. 501217485
Encompass_Preferred
Formula: C11H7KN2O3S2, Molecular weight: 318.42 g/mol, Store at -15°C, Synonyms: (S)-4,5-Dihydro-2-(6-hydroxy-2-benzothiazolyl)-4-thiazolecarboxylic acid potassium salt.
Catalog No. 50-121-7485 Supplier Carbosynth LLC. Supplier No. L82200.25G
Formula: C11H7KN2O3S2, Molecular weight: 318.42 g/mol, Store at -15°C, Synonyms: (S)-4,5-Dihydro-2-(6-hydroxy-2-benzothiazolyl)-4-thiazolecarboxylic acid potassium salt.