Learn More
Abcam D-Luciferin, 6'-methyl ether, 5MG
Catalog No. 50996083
Encompass_Preferred
MW 294.3 g/mol, Purity >95%. Firefly luciferase inhibitor. Lucifern analog. Useful in luciferase / cytochrome P450 dual assay systems. Dealkylase removes the methyl ether to release a luciferase substrate which produces light in direct proportion to cytochrome P450 activity.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-996-083 Supplier Abcam Supplier No. AB2753695MG
Add to Cart
Add to Cart
MW 294.3 g/mol, Purity >95%. Firefly luciferase inhibitor. Lucifern analog. Useful in luciferase / cytochrome P450 dual assay systems. Dealkylase removes the methyl ether to release a luciferase substrate which produces light in direct proportion to cytochrome P450 activity.
The product is subject to the following: Abcam Restricted Use Statement