Learn More
Carbosynth LLC. D-Luciferin Firefly, sodium salt monohydrate, 103404-75-7, MFCD00044938, 1 g
SDP
Catalog No. 50853140
Encompass_Preferred
Formula: C11H9N2NaO4S2, Molecular weight: 320.32 g/mol, 98%, Store at -15°C, Synonyms: (S)-4,5-Dihydro-2-(6-hydroxy-2-benzothiazolyl)-4-thiazolecarboxylic acid sodium salt.
Catalog No. 50-853-140 Supplier Carbosynth LLC. Supplier No. FL086091G
Formula: C11H9N2NaO4S2, Molecular weight: 320.32 g/mol, 98%, Store at -15°C, Synonyms: (S)-4,5-Dihydro-2-(6-hydroxy-2-benzothiazolyl)-4-thiazolecarboxylic acid sodium salt.