Learn More
Aat Bioquest D-Luciferin phosphate *CAS 145613-12-3*
SDP
Catalog No. 501953394
Encompass_Preferred
D-Luciferin phosphate is a highly sensitive bioluminescent substrate for detecting alkaline phosphatases and protein phosphatases by luminometric detection. It is used in combination with luciferase/ATP system.
Catalog No. 50-195-3394 Supplier Aat Bioquest Supplier No. 12512
D-Luciferin phosphate is a highly sensitive bioluminescent substrate for detecting alkaline phosphatases and protein phosphatases by luminometric detection. It is used in combination with luciferase/ATP system.