Learn More
TARGETMOL CHEMICALS INC D-Luciferin potassium 100MG
SDP
Catalog No. 502879015
Encompass_Preferred
Also available in 1 g, 10 mg, 50 mg, 500 mg and bulk. Please contact Fisher for quotes. D-Luciferin potassium (D-Luciferin K Salt) is a popular bioluminescent substrate of luciferase in the presence of ATP. Purity 99.13%
Catalog No. 50-287-9015 Supplier TARGETMOL CHEMICALS INC Supplier No. T4139100MG
Also available in 1 g, 10 mg, 50 mg, 500 mg and bulk. Please contact Fisher for quotes. D-Luciferin potassium (D-Luciferin K Salt) is a popular bioluminescent substrate of luciferase in the presence of ATP. Purity 99.13%