Learn More
Medchemexpress LLC D-luciferin potassium | 115144-35-9 | 100.0% | 318.41 | C11H7KN2O3S2 | 250MG
SDP
Catalog No. 5000238164
Encompass_Preferred
D-Luciferin potassium is the potassium salt of D-luciferin, the natural substrate for firefly luciferase. In the presence of ATP and appropriate cofactors it undergoes enzymatic oxidation to produce rapid, proportional chemiluminescent light, making it suitable for reporter gene assays, cell viability measurements, and sensitive ATP detection. Supplied as a high-purity solid and available in multiple pack sizes for research use.
  • High-purity potassium salt for reliable luminescent assays.
  • Produces rapid, proportional chemiluminescent signal.
  • Compatible with luciferase-based reporter and ATP assays.
  • Supplied as a solid, stable form for storage and handling.
  • Available in a range of pack sizes to suit experimental needs.
Catalog No. 50-002-38164 Supplier Medchemexpress LLC Supplier No. HY12591B250MG
D-Luciferin potassium is the potassium salt of D-luciferin, the natural substrate for firefly luciferase. In the presence of ATP and appropriate cofactors it undergoes enzymatic oxidation to produce rapid, proportional chemiluminescent light, making it suitable for reporter gene assays, cell viability measurements, and sensitive ATP detection. Supplied as a high-purity solid and available in multiple pack sizes for research use.
  • High-purity potassium salt for reliable luminescent assays.
  • Produces rapid, proportional chemiluminescent signal.
  • Compatible with luciferase-based reporter and ATP assays.
  • Supplied as a solid, stable form for storage and handling.
  • Available in a range of pack sizes to suit experimental needs.