Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-luciferin potassium | 115144-35-9 | 100.0% | 318.41 g·mol^-1 | C11H7KN2O3S2 | 1G
SDP

Catalog No. 5000234996
Encompass_Preferred

D-Luciferin potassium is the water-soluble potassium salt of D-luciferin, the natural substrate for firefly luciferase. It produces yellow-green bioluminescence upon ATP-dependent oxidation and is used for biochemical luciferase assays, cell-based reporter assays, and in vivo imaging. The compound is supplied as a solid powder suitable for preparation of aqueous solutions for experimental use.

  • Water-soluble potassium salt for easy aqueous formulation.
  • Suitable for bioluminescent luciferase assays and imaging.
  • High analytical purity (manufacturer lists 99.97%).
  • Molecular weight 318.41 g·mol⁻¹ and formula C11H7KN2O3S2.
  • Available in multiple pack sizes including 1 g for laboratory use.

Catalog No. 50-002-34996 Supplier Medchemexpress LLC Supplier No. HY12591B1G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-Luciferin potassium is the water-soluble potassium salt of D-luciferin, the natural substrate for firefly luciferase. It produces yellow-green bioluminescence upon ATP-dependent oxidation and is used for biochemical luciferase assays, cell-based reporter assays, and in vivo imaging. The compound is supplied as a solid powder suitable for preparation of aqueous solutions for experimental use.

  • Water-soluble potassium salt for easy aqueous formulation.
  • Suitable for bioluminescent luciferase assays and imaging.
  • High analytical purity (manufacturer lists 99.97%).
  • Molecular weight 318.41 g·mol⁻¹ and formula C11H7KN2O3S2.
  • Available in multiple pack sizes including 1 g for laboratory use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.