Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-luciferin potassium | 115144-35-9 | 99.97% | 318.41 g/mol | C11H7KN2O3S2 | 10MM 1ML
SDP

Catalog No. 5000236511
Encompass_Preferred

D-Luciferin potassium is the potassium salt of D-luciferin, the natural substrate for firefly luciferase. In the presence of luciferase and ATP it produces a yellow-green bioluminescent signal (peak ~560 nm) used for luciferase assays and bioluminescence imaging. Supplied as a 10 mM ready-to-use solution (1 mL), it provides high purity and convenient handling for assay workflows.

  • High purity: 99.97% by supplier analysis.
  • Ready-to-use 10 mM stock for rapid assay setup.
  • Generates strong bioluminescent signal with peak emission near 560 nm.
  • Soluble in water (≈54.6 mM) with warming and sonication as needed.
  • Suitable for luciferase reporter assays and bioluminescence imaging applications.
  • Storage: sealed, protected from moisture and light; in solution store at -80°C for long-term stability.

Catalog No. 50-002-36511 Supplier Medchemexpress LLC Supplier No. HY12591B10MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-Luciferin potassium is the potassium salt of D-luciferin, the natural substrate for firefly luciferase. In the presence of luciferase and ATP it produces a yellow-green bioluminescent signal (peak ~560 nm) used for luciferase assays and bioluminescence imaging. Supplied as a 10 mM ready-to-use solution (1 mL), it provides high purity and convenient handling for assay workflows.

  • High purity: 99.97% by supplier analysis.
  • Ready-to-use 10 mM stock for rapid assay setup.
  • Generates strong bioluminescent signal with peak emission near 560 nm.
  • Soluble in water (≈54.6 mM) with warming and sonication as needed.
  • Suitable for luciferase reporter assays and bioluminescence imaging applications.
  • Storage: sealed, protected from moisture and light; in solution store at -80°C for long-term stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.