Learn More
Medchemexpress LLC D-luciferin (potassium) | 115144-35-9 | 99.1% | 318.41 g/mol | C11H7KN2O3S2 | 500MG
SDP
Catalog No. 5000239325
Encompass_Preferred
D-Luciferin potassium is the potassium salt of D-luciferin, the native substrate for firefly luciferase. It is provided as a water-soluble reagent for use in in vitro and in vivo bioluminescence assays and imaging, intended to deliver consistent luminescent signals with appropriate storage for stability.
  • Water-soluble potassium salt form suitable for aqueous assays.
  • Natural substrate for luciferase-based bioluminescent detection.
  • High purity (99.1% by LCMS) for reproducible assay results.
  • Molecular weight 318.41 g/mol enables precise dosing.
  • Stable when stored sealed at 4°C; in solvent stable at -80°C for up to 2 years.
  • Suitable for reporter assays and in vivo bioluminescence imaging.
Catalog No. 50-002-39325 Supplier Medchemexpress LLC Supplier No. HY12591B500MG
D-Luciferin potassium is the potassium salt of D-luciferin, the native substrate for firefly luciferase. It is provided as a water-soluble reagent for use in in vitro and in vivo bioluminescence assays and imaging, intended to deliver consistent luminescent signals with appropriate storage for stability.
  • Water-soluble potassium salt form suitable for aqueous assays.
  • Natural substrate for luciferase-based bioluminescent detection.
  • High purity (99.1% by LCMS) for reproducible assay results.
  • Molecular weight 318.41 g/mol enables precise dosing.
  • Stable when stored sealed at 4°C; in solvent stable at -80°C for up to 2 years.
  • Suitable for reporter assays and in vivo bioluminescence imaging.