Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-luciferin potassium | 115144-35-9 | MFCD00044928 | 99.97% | 318.41 | C11H7KN2O3S2 | 100MG
SDP

Catalog No. 5000236510
Encompass_Preferred

D-luciferin potassium is the water-soluble potassium salt of D-luciferin, the natural substrate for firefly luciferase. It is provided as a solid for preparation of aqueous solutions used in both in vitro and in vivo bioluminescent assays and imaging applications.

  • Water-soluble potassium salt of D-luciferin.
  • Serves as substrate for firefly luciferase in bioluminescent assays.
  • High purity suitable for research applications (reported 99.97%).
  • Applicable to both in vitro assays and in vivo imaging workflows.
  • Soluble in aqueous buffers for easy preparation of working solutions.
  • Molecular weight 318.41; chemical formula C11H7KN2O3S2.

Catalog No. 50-002-36510 Supplier Medchemexpress LLC Supplier No. HY12591B100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-luciferin potassium is the water-soluble potassium salt of D-luciferin, the natural substrate for firefly luciferase. It is provided as a solid for preparation of aqueous solutions used in both in vitro and in vivo bioluminescent assays and imaging applications.

  • Water-soluble potassium salt of D-luciferin.
  • Serves as substrate for firefly luciferase in bioluminescent assays.
  • High purity suitable for research applications (reported 99.97%).
  • Applicable to both in vitro assays and in vivo imaging workflows.
  • Soluble in aqueous buffers for easy preparation of working solutions.
  • Molecular weight 318.41; chemical formula C11H7KN2O3S2.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.