Learn More
Gold Biotechnology Inc D Luciferin Potassium 500mg
SDP
Catalog No. 5000562099
Encompass_Preferred
L uciferin is a common bioluminescent reporter used for in vivo imaging of the expression of luciferase This water soluble substrate for the firefly luciferase enzyme utilizes ATP and Mg2 as cofactors to emit a characteristic yellow-green emission in the presence of oxygen which shifts to red light in vivo at 37 C Through the utilization of ATP the reaction can be further used to indicate the presence of energy or life in order to function as a life-death stain
Catalog No. 50-005-62099 Supplier Gold Biotechnology Inc Supplier No. LUCK500
L uciferin is a common bioluminescent reporter used for in vivo imaging of the expression of luciferase This water soluble substrate for the firefly luciferase enzyme utilizes ATP and Mg2 as cofactors to emit a characteristic yellow-green emission in the presence of oxygen which shifts to red light in vivo at 37 C Through the utilization of ATP the reaction can be further used to indicate the presence of energy or life in order to function as a life-death stain