Learn More
Medchemexpress LLC D-luciferin potassium salt | 115144-35-9 | MFCD00044928 | 100.0% | 318.41 | C11H7KN2O3S2 | 5MG
SDP
Catalog No. 5000238305
Encompass_Preferred
D-luciferin potassium is the potassium salt form of D-luciferin, the natural substrate for firefly luciferase. Supplied as a high-purity solid, it is used as a chemiluminescent substrate in biochemical assays and in vivo and in vitro bioluminescence imaging. The potassium salt improves aqueous solubility, facilitating assay preparation and reagent formulation.
  • High purity suitable for biochemical assays and imaging.
  • Potassium salt form enhances water solubility for easy dissolution.
  • Produces ATP-dependent bioluminescence with luciferase enzymes.
  • Available in small scales for research and formulation development.
  • Provided with product datasheet and safety data for laboratory use.
Catalog No. 50-002-38305 Supplier Medchemexpress LLC Supplier No. HY12591B5MG
D-luciferin potassium is the potassium salt form of D-luciferin, the natural substrate for firefly luciferase. Supplied as a high-purity solid, it is used as a chemiluminescent substrate in biochemical assays and in vivo and in vitro bioluminescence imaging. The potassium salt improves aqueous solubility, facilitating assay preparation and reagent formulation.
  • High purity suitable for biochemical assays and imaging.
  • Potassium salt form enhances water solubility for easy dissolution.
  • Produces ATP-dependent bioluminescence with luciferase enzymes.
  • Available in small scales for research and formulation development.
  • Provided with product datasheet and safety data for laboratory use.