Learn More
Biotium D-Luciferin, POTASSIUM SALT
Catalog No. NC1123013
Encompass_Preferred
D-Luciferin is a substrate for the enzyme firefly luciferase. Chemiluminescence at 560 nm is generated when luciferin is oxidized by an ATP-dependent process, catalyzed by luciferase. Luciferase is encoded by the luc gene, which is widely used as a reporter gene in a variety of cells. Because of the intrinsic low background of chemiluminescence technique, detection of the luc gene expression can be made at very low level. In addition, luciferin/luciferase has been used to measure 1015 molar quantity of ATP. D-luciferin, potassium salt is a water-soluble form of D-luciferin.Properties: Light yellow solid soluble in water; Store at -20°C and protect from light; C11H7KN2O3S2; MW 318.4; CAS No. [115144-35-9].
Catalog No. NC1123013 Supplier Biotium Supplier No. 101012
D-Luciferin is a substrate for the enzyme firefly luciferase. Chemiluminescence at 560 nm is generated when luciferin is oxidized by an ATP-dependent process, catalyzed by luciferase. Luciferase is encoded by the luc gene, which is widely used as a reporter gene in a variety of cells. Because of the intrinsic low background of chemiluminescence technique, detection of the luc gene expression can be made at very low level. In addition, luciferin/luciferase has been used to measure 1015 molar quantity of ATP. D-luciferin, potassium salt is a water-soluble form of D-luciferin.Properties: Light yellow solid soluble in water; Store at -20°C and protect from light; C11H7KN2O3S2; MW 318.4; CAS No. [115144-35-9].