Learn More
Medchemexpress LLC HY-12591B 50mg Medchemexpress, D-Luciferin potassium salt CAS:115144-35-9 Purity:98%
SDP
Catalog No. 501637780
Encompass_Preferred
Medchemexpress,HY-12591B 50mg D-Luciferin potassium salt CAS:115144-35-9 Formula:C11H7KN2O3S2 D-Luciferin potassium salt is the substrate of luciferases that catalyze the production of light in bioluminescent insects. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-163-7780 Supplier Medchemexpress LLC Supplier No. HY12591B50MG
Medchemexpress,HY-12591B 50mg D-Luciferin potassium salt CAS:115144-35-9 Formula:C11H7KN2O3S2 D-Luciferin potassium salt is the substrate of luciferases that catalyze the production of light in bioluminescent insects. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.