Learn More
Apexbio Technology LLC D-Luciferin (potassium salt) 115144-35-9 1g
SDP
Supplier:  Apexbio Technology LLC C36541000
Encompass_Preferred
D-Luciferin (potassium salt) a potassium salt derivative of D-luciferin serves as the bioluminescent substrate for firefly luciferase The enzyme-mediated oxidation of D-luciferin in the presence of ATP and oxygen produces bioluminescent emission at approximately 560 nm without the need for external excitation This property is applied extensively in bioluminescence imaging (BLI) experiments including live-cell and small-animal imaging Cells genetically engineered to express luciferase utilize D-luciferin to generate optical signals facilitating non-invasive visualization and tracking of biological events such as tumor growth gene expression dynamics in vivo distribution and therapeutic responses
Catalog No. 50-000-64295