Learn More
Cayman Chemical D-Luciferin (potassium salt), 115144-35-9, 1 g
SDP
Supplier:  Cayman Chemical 146811
Encompass_Preferred
A chemiluminescent substrate of firefly luciferase which produces light upon oxidative decarboxylation in the presence of ATP
Catalog No. NC1425692