Learn More
Bioworld D-Luciferin, Potassium Salt, 100 mg
Catalog No. 50255853
Encompass_Preferred
Produces bioluminescence when oxidized by firefly luciferase. Allows highly sensitive bioluminescent measurement. Specification:Water Soluble FormL-Luciferin: <0.5%Protect from light and mositureAppearance: Solid
Catalog No. 50-255-853 Supplier Bioworld Supplier No. 404000762
Produces bioluminescence when oxidized by firefly luciferase. Allows highly sensitive bioluminescent measurement. Specification:Water Soluble FormL-Luciferin: <0.5%Protect from light and mositureAppearance: Solid