Learn More
Bioworld D-Luciferin, Potassium Salt, 1 g
Catalog No. 50255855
Encompass_Preferred
Produces bioluminescence when oxidized by firefly luciferase. Allows highly sensitive bioluminescent measurement. Specification:Water Soluble FormL-Luciferin: <0.5%Protect from light and mositureAppearance: Solid
Catalog No. 50-255-855 Supplier Bioworld Supplier No. 404000764
Produces bioluminescence when oxidized by firefly luciferase. Allows highly sensitive bioluminescent measurement. Specification:Water Soluble FormL-Luciferin: <0.5%Protect from light and mositureAppearance: Solid