Learn More
Abcam D-Luciferin potassium salt, chemiluminescent luciferase substrate, 100MG
Catalog No. 502204934
Encompass_Preferred
MW 318.4 Da, Purity >98%. Chemiluminescent luciferase substrate. Water soluble derivative of D-Luciferin (ab143654) and analog of D-Luciferin sodium salt (ab145164). Emits light on oxidative decarboxylation in the presence of ATP providing a bioluminescent signal for cellular ATP levels in vivo..
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-220-4934 Supplier Abcam Supplier No. AB143655100MG
MW 318.4 Da, Purity >98%. Chemiluminescent luciferase substrate. Water soluble derivative of D-Luciferin (ab143654) and analog of D-Luciferin sodium salt (ab145164). Emits light on oxidative decarboxylation in the presence of ATP providing a bioluminescent signal for cellular ATP levels in vivo..
The product is subject to the following: Abcam Restricted Use Statement