Learn More
AnaSpec D - Luciferin, potassium salt *UltraPure Grade*, 25mg
Catalog No. 50850171
Encompass
D-Luciferin, potassium salt *UltraPure Grade*, ; MW 318.41; (25 mg) Luminescent substrate for firefly luciferase.
Catalog No. 50-850-171 Supplier AnaSpec Supplier No. AS82252
D-Luciferin, potassium salt *UltraPure Grade*, ; MW 318.41; (25 mg) Luminescent substrate for firefly luciferase.