Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD D- Luciferin Potassium Salt

Catalog No. BDB556888 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
250 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB556888 250 mg
BDB556878 50 mg
BDB556879 100 mg
BDB556877 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. BDB556888 Supplier BD Supplier No. 556888
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Synthetically made substrate which can be catalyzed by firefly luciferase

  • Also known as beetle luciferin
  • Lyophilized in form of monopotassium salt
  • Purified by HPLC to remove dehydroluciferin and L-Luciferin
  • Form dehydroluciferin in presence of ultraviolet light and/or moisture

Luminometer

TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized
Type D-Luciferin
Quantity 250 mg
Content And Storage -20°C
For Use With (Application) Luminometer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.