Learn More
TARGETMOL CHEMICALS INC D-Luciferin Sodium 100MG
SDP
Catalog No. 502878372
Encompass_Preferred
Also available in 1 mL, 10 mg, 25 mg, 50 mg, 200 mg, 500 mg and bulk. Please contact Fisher for quotes. D-Luciferin Sodium is a heterocyclic light-emitting compound and natural ligand for luciferase used to detect cell activity. The salt form of luciferin dissolves in water or other typical buffers. Purity 99.65%
Catalog No. 50-287-8372 Supplier TARGETMOL CHEMICALS INC Supplier No. T19743100MG
Also available in 1 mL, 10 mg, 25 mg, 50 mg, 200 mg, 500 mg and bulk. Please contact Fisher for quotes. D-Luciferin Sodium is a heterocyclic light-emitting compound and natural ligand for luciferase used to detect cell activity. The salt form of luciferin dissolves in water or other typical buffers. Purity 99.65%