Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-luciferin sodium | 103404-75-7 | 99.9% | 302.30 | C11H7N2NaO3S2 | 1G
SDP

Catalog No. 5000235331
Encompass_Preferred

D-Luciferin sodium is the water-soluble sodium salt of D-luciferin, the natural substrate for firefly luciferase. Upon enzymatic oxidation it produces rapid yellow-green chemiluminescence (peak ~560 nm) used in reporter gene assays, ATP detection, and bioluminescence imaging. Supplied as a solid with high purity and multiple pack sizes, it should be stored protected from moisture and light.

  • High purity (99.92%) for consistent assay results.
  • Water-soluble sodium salt for easy preparation of solutions.
  • Rapid yellow-green chemiluminescence with peak emission at ~560 nm.
  • Suitable for reporter gene assays, ATP quantification, and imaging applications.
  • Available in multiple pack sizes including 1 g.
  • Stable when stored sealed at 4°C away from moisture and light.

Catalog No. 50-002-35331 Supplier Medchemexpress LLC Supplier No. HY125911G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-Luciferin sodium is the water-soluble sodium salt of D-luciferin, the natural substrate for firefly luciferase. Upon enzymatic oxidation it produces rapid yellow-green chemiluminescence (peak ~560 nm) used in reporter gene assays, ATP detection, and bioluminescence imaging. Supplied as a solid with high purity and multiple pack sizes, it should be stored protected from moisture and light.

  • High purity (99.92%) for consistent assay results.
  • Water-soluble sodium salt for easy preparation of solutions.
  • Rapid yellow-green chemiluminescence with peak emission at ~560 nm.
  • Suitable for reporter gene assays, ATP quantification, and imaging applications.
  • Available in multiple pack sizes including 1 g.
  • Stable when stored sealed at 4°C away from moisture and light.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.