Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-luciferin (sodium) | 103404-75-7 | 99.9% | 302.30 | C11H7N2NaO3S2 | 500MG
SDP

Catalog No. 5000235613
Encompass_Preferred

D-Luciferin sodium is the sodium salt of D-luciferin, the natural substrate for firefly luciferase used to generate bioluminescent signals in reporter assays and imaging. Supplied as a light yellow solid with high purity, it is suitable for in vitro and in vivo bioluminescence applications.

  • Natural substrate for firefly luciferase.
  • Produces bioluminescent signal for reporter assays and imaging.
  • High purity (99.9%).
  • Molecular weight 302.30.
  • Formula C11H7N2NaO3S2.
  • Available in multiple pack sizes, including 500 MG.
  • Solid, light yellow to yellow appearance.
  • Store sealed, away from moisture and light; in solvent store at -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-35613 Supplier Medchemexpress LLC Supplier No. HY12591500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-Luciferin sodium is the sodium salt of D-luciferin, the natural substrate for firefly luciferase used to generate bioluminescent signals in reporter assays and imaging. Supplied as a light yellow solid with high purity, it is suitable for in vitro and in vivo bioluminescence applications.

  • Natural substrate for firefly luciferase.
  • Produces bioluminescent signal for reporter assays and imaging.
  • High purity (99.9%).
  • Molecular weight 302.30.
  • Formula C11H7N2NaO3S2.
  • Available in multiple pack sizes, including 500 MG.
  • Solid, light yellow to yellow appearance.
  • Store sealed, away from moisture and light; in solvent store at -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.