Learn More
Medchemexpress LLC D-luciferin sodium | 103404-75-7 | 99.9% | 302.30 g·mol⁻1 | C11H7N2NaO3S2 | 10MG
SDP
Catalog No. 5000235931
Encompass_Preferred
D-Luciferin sodium is the sodium salt of D-luciferin, the natural substrate for luciferase enzymes. It is supplied as a high-purity reagent for sensitive bioluminescent detection in applications such as reporter gene assays, cell viability testing, and in vivo imaging, providing a reliable luminescent signal when used with appropriate luciferase systems.
  • High purity suitable for sensitive bioluminescent assays.
  • Water-soluble sodium salt formulation for easy preparation.
  • Compatible with in vitro and in vivo luciferase-based imaging.
  • Available in multiple pack sizes for small- and large-scale experiments.
  • Stable when stored according to supplier instructions (refer to SDS).
Catalog No. 50-002-35931 Supplier Medchemexpress LLC Supplier No. HY1259110MG
D-Luciferin sodium is the sodium salt of D-luciferin, the natural substrate for luciferase enzymes. It is supplied as a high-purity reagent for sensitive bioluminescent detection in applications such as reporter gene assays, cell viability testing, and in vivo imaging, providing a reliable luminescent signal when used with appropriate luciferase systems.
  • High purity suitable for sensitive bioluminescent assays.
  • Water-soluble sodium salt formulation for easy preparation.
  • Compatible with in vitro and in vivo luciferase-based imaging.
  • Available in multiple pack sizes for small- and large-scale experiments.
  • Stable when stored according to supplier instructions (refer to SDS).