Learn More
Medchemexpress LLC D-luciferin sodium | 103404-75-7 | MFCD00044938 | >95.0% | 302.3 g/mol | C11H7N2NaO3S2 | 100MG
SDP
Catalog No. 5000235567
Encompass_Preferred
D-luciferin sodium is the water-soluble sodium salt of firefly luciferin used as a chemiluminescent substrate for luciferase assays in vitro and in vivo. Supplied as a powder, it dissolves readily in aqueous or organic solvents to prepare assay solutions for reporter gene assays, bioluminescence imaging, and ATP-dependent measurements.
  • Water-soluble sodium salt suitable for luciferase-based bioluminescence assays.
  • High purity (>95%) for reliable assay performance.
  • Soluble in water and DMSO (solubility reported to ~25 mM) for easy preparation.
  • Provided as a lyophilized powder for accurate dosing and storage.
  • Stable when stored desiccated at -20°C and protected from light.
Catalog No. 50-002-35567 Supplier Medchemexpress LLC Supplier No. HY12591100MG
D-luciferin sodium is the water-soluble sodium salt of firefly luciferin used as a chemiluminescent substrate for luciferase assays in vitro and in vivo. Supplied as a powder, it dissolves readily in aqueous or organic solvents to prepare assay solutions for reporter gene assays, bioluminescence imaging, and ATP-dependent measurements.
  • Water-soluble sodium salt suitable for luciferase-based bioluminescence assays.
  • High purity (>95%) for reliable assay performance.
  • Soluble in water and DMSO (solubility reported to ~25 mM) for easy preparation.
  • Provided as a lyophilized powder for accurate dosing and storage.
  • Stable when stored desiccated at -20°C and protected from light.