Learn More
Medchemexpress LLC D-luciferin sodium | 103404-75-7 | MFCD00044938 | 99.9% | 302.30 | C11H7N2NaO3S2 | 5MG
SDP
Catalog No. 5000235329
Encompass_Preferred
D-Luciferin sodium is the sodium salt of D-luciferin, a bioluminescent substrate for firefly luciferase. Upon oxidation in the presence of luciferase, ATP, and oxygen it produces a yellow-green light (peak ~560 nm), making it suitable for sensitive reporter assays and bioluminescence imaging. Supplied as a light-yellow solid with high purity and a defined molecular weight for consistent assay performance.
  • High purity for reliable assay results.
  • Generates a strong yellow-green bioluminescent signal (peak ~560 nm).
  • Suitable for in vitro and in vivo luciferase reporter assays.
  • Sodium salt form for good aqueous solubility and easy preparation.
  • Supplied in small, accurately weighed quantities for precise dosing.
Catalog No. 50-002-35329 Supplier Medchemexpress LLC Supplier No. HY125915MG
D-Luciferin sodium is the sodium salt of D-luciferin, a bioluminescent substrate for firefly luciferase. Upon oxidation in the presence of luciferase, ATP, and oxygen it produces a yellow-green light (peak ~560 nm), making it suitable for sensitive reporter assays and bioluminescence imaging. Supplied as a light-yellow solid with high purity and a defined molecular weight for consistent assay performance.
  • High purity for reliable assay results.
  • Generates a strong yellow-green bioluminescent signal (peak ~560 nm).
  • Suitable for in vitro and in vivo luciferase reporter assays.
  • Sodium salt form for good aqueous solubility and easy preparation.
  • Supplied in small, accurately weighed quantities for precise dosing.