Learn More
Medchemexpress LLC D-luciferin sodium salt | 103404-75-7 | 99.9% | 302.30 g/mol | C11H7N2NaO3S2 | 200MG
SDP
Catalog No. 5000236588
Encompass_Preferred
D-Luciferin sodium is the sodium salt of D-luciferin, the natural substrate for firefly luciferase. Upon oxidation by luciferase it produces bioluminescence, making it suitable for in vitro and in vivo imaging and quantitative luciferase assays. The material is supplied as a solid for preparation of aqueous stock solutions and experimental use.
  • High purity (~99.9%).
  • Water-soluble sodium salt form suitable for biological assays.
  • Enables bioluminescent imaging and quantitative luciferase assays.
  • Molecular weight 302.30 g/mol.
  • Available in multiple pack sizes from 5 mg to 50 g.
Catalog No. 50-002-36588 Supplier Medchemexpress LLC Supplier No. HY12591200MG
D-Luciferin sodium is the sodium salt of D-luciferin, the natural substrate for firefly luciferase. Upon oxidation by luciferase it produces bioluminescence, making it suitable for in vitro and in vivo imaging and quantitative luciferase assays. The material is supplied as a solid for preparation of aqueous stock solutions and experimental use.
  • High purity (~99.9%).
  • Water-soluble sodium salt form suitable for biological assays.
  • Enables bioluminescent imaging and quantitative luciferase assays.
  • Molecular weight 302.30 g/mol.
  • Available in multiple pack sizes from 5 mg to 50 g.