Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-luciferin sodium salt | 103404-75-7 | MFCD00044938 | 99.9% | 302.3 g/mol | C11H7N2NaO3S2 | 5G
SDP

Catalog No. 5000235612
Encompass_Preferred

D-luciferin sodium is the water-soluble sodium salt of firefly luciferin used as a chemiluminescent substrate for luciferase-based assays and imaging. It supplies a reliable light-emitting substrate for quantitative enzymatic assays and bioluminescent imaging applications.

  • Water-soluble sodium salt for easy assay preparation.
  • Suitable as a chemiluminescent substrate for luciferase-based assays and imaging.
  • High purity (99.9%) for reproducible results.
  • Soluble in water for preparation of assay solutions.
  • Stable as a solid when stored sealed, away from moisture and light.
  • Solution storage: -80°C up to 6 months or -20°C up to 1 month.
  • Molecular weight 302.3 g/mol; formula C11H7N2NaO3S2.
  • Available in lab-scale pack sizes including 5 g.

Catalog No. 50-002-35612 Supplier Medchemexpress LLC Supplier No. HY125915G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-luciferin sodium is the water-soluble sodium salt of firefly luciferin used as a chemiluminescent substrate for luciferase-based assays and imaging. It supplies a reliable light-emitting substrate for quantitative enzymatic assays and bioluminescent imaging applications.

  • Water-soluble sodium salt for easy assay preparation.
  • Suitable as a chemiluminescent substrate for luciferase-based assays and imaging.
  • High purity (99.9%) for reproducible results.
  • Soluble in water for preparation of assay solutions.
  • Stable as a solid when stored sealed, away from moisture and light.
  • Solution storage: -80°C up to 6 months or -20°C up to 1 month.
  • Molecular weight 302.3 g/mol; formula C11H7N2NaO3S2.
  • Available in lab-scale pack sizes including 5 g.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.