Learn More
Apexbio Technology LLC D-Luciferin sodium salt(Synonyms: Sodium D-Luciferin, Luciferin sodium salt, Luciferase substrate), 10mg, CAS: 103404-75-7.
SDP
Catalog No. 5000060701
Encompass_Preferred
D-Luciferin sodium salt (CAS 103404-75-7) is a bioluminescent substrate widely utilized in biomedical research and diagnostic applications Upon entering cells D-Luciferin undergoes enzymatic oxidation catalyzed by luciferase enzymes a reaction requiring ATP and molecular oxygen that emits detectable light This ATP-dependent bioluminescent reaction enables non-invasive monitoring of intracellular metabolic status and energy availability Commonly employed in bioluminescence imaging D-Luciferin sodium salt serves as a powerful tool to assess cellular viability metabolism and gene expression in research investigating cell biology oncology and metabolism
Catalog No. 50-000-60701 Supplier Apexbio Technology LLC Supplier No. B831110
D-Luciferin sodium salt (CAS 103404-75-7) is a bioluminescent substrate widely utilized in biomedical research and diagnostic applications Upon entering cells D-Luciferin undergoes enzymatic oxidation catalyzed by luciferase enzymes a reaction requiring ATP and molecular oxygen that emits detectable light This ATP-dependent bioluminescent reaction enables non-invasive monitoring of intracellular metabolic status and energy availability Commonly employed in bioluminescence imaging D-Luciferin sodium salt serves as a powerful tool to assess cellular viability metabolism and gene expression in research investigating cell biology oncology and metabolism