Learn More
Gold Biotechnology Inc D-Luciferin, Sodium Salt (Proven and Published™) 1 g
SDP
Catalog No. NC1972377
Encompass_Preferred
Luciferin is a common bioluminescent reporter used for in vivo imaging of the expression of luciferase. This water soluble substrate for the firefly luciferase enzyme utilizes ATP and Mg2+ as cofactors to emit a characteristic yellow-green emission in the presence of oxygen, which shifts to red light in vivo at 37°C. Through the utilization of ATP, the reaction can be further used to indicate the presence of energy or life in order to function as a life-death stain.
Catalog No. NC1972377 Supplier Gold Biotechnology Inc Supplier No. LUCANA1G
Luciferin is a common bioluminescent reporter used for in vivo imaging of the expression of luciferase. This water soluble substrate for the firefly luciferase enzyme utilizes ATP and Mg2+ as cofactors to emit a characteristic yellow-green emission in the presence of oxygen, which shifts to red light in vivo at 37°C. Through the utilization of ATP, the reaction can be further used to indicate the presence of energy or life in order to function as a life-death stain.