Learn More
Selleck Chemical LLC D-Luciferin sodium salt
SDP
Catalog No. 5000002618
Encompass_Preferred
D-Luciferin (D-(-)-Luciferin Firefly luciferin) sodium salt is the natural substrate of luciferases that catalyze the production of light in bioluminescent insects
Catalog No. 50-000-02618 Supplier Selleck Chemical LLC Supplier No. S692425MG
D-Luciferin (D-(-)-Luciferin Firefly luciferin) sodium salt is the natural substrate of luciferases that catalyze the production of light in bioluminescent insects