Learn More
Alkali Scientific D Luciferin Sodium Salt [4,5 Dihydro 2 (6 hydroxy 2 benzothiazolyl) 4 thia zolecarboxylic acid, sodium salt], 100 Mg
SDP
Catalog No. 5000659814
Encompass_Preferred
D-Luciferin Sodium Salt is a key substrate used in bioluminescence assays, particularly for detecting the activity of luciferase in biological systems. This reagent, in its sodium salt form, is used in laboratory applications to measure light emission through a reaction with the enzyme luciferase. The product is commonly used in applications such as gene expression studies, cell viability assays, and ATP detection. With its ability to generate light in the presence of luciferase, it provides an effective and highly sensitive method for biochemical assays. This 100mg bottle is ideal for laboratory-scale experimentation and analysis.
Catalog No. 50-006-59814 Supplier Alkali Scientific Supplier No. DLU004CP
D-Luciferin Sodium Salt is a key substrate used in bioluminescence assays, particularly for detecting the activity of luciferase in biological systems. This reagent, in its sodium salt form, is used in laboratory applications to measure light emission through a reaction with the enzyme luciferase. The product is commonly used in applications such as gene expression studies, cell viability assays, and ATP detection. With its ability to generate light in the presence of luciferase, it provides an effective and highly sensitive method for biochemical assays. This 100mg bottle is ideal for laboratory-scale experimentation and analysis.