Learn More
Abcam D-Luciferin sodium salt, Chemiluminescent luciferase substrate, 100MG
Catalog No. 502204938
Encompass_Preferred
MW 302.3 g/mol, Purity >95%. Chemiluminescent luciferase substrate. Water soluble derivative of D-Luciferin (ab143654) and analog of D-Luciferin potassium salt (ab143655). Excitation is pH dependent (pH exλ = 327 and 385 nm for pH 4 and pH 11 respectively), emission is pH independent. Emits light on oxidative decarboxylation in the presence of ATP providing a bioluminescent signal for cellular ATP levels in vivo..
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-220-4938 Supplier Abcam Supplier No. AB145164100MG
MW 302.3 g/mol, Purity >95%. Chemiluminescent luciferase substrate. Water soluble derivative of D-Luciferin (ab143654) and analog of D-Luciferin potassium salt (ab143655). Excitation is pH dependent (pH exλ = 327 and 385 nm for pH 4 and pH 11 respectively), emission is pH independent. Emits light on oxidative decarboxylation in the presence of ATP providing a bioluminescent signal for cellular ATP levels in vivo..
The product is subject to the following: Abcam Restricted Use Statement